missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-chlorophenyltrifluoroborate, 96%

Manufacturer:  Alfa Aesar™ H5323106

 View more versions of this product

Catalog No. AAH5323106



Description

Description

Applications
Potassium 4-chlorophenyltrifluoroborate is involved in Suzuki Cross-Coupling reaction as potent boronic acid surrogates.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents and acids. Store at room temperature.
Specifications

Specifications

Potassium 4-chlorophenyltrifluoroborate
5g
C6H4BClF3K
potassium 4-chlorophenyltrifluoroborate, potassium 4-chlorophenyl trifluoroborate, potassium 4-chlorophenyl trifluoroboranuide, pubchem11348, potassium 4-chlorophenyltrifluoroborate, potassium 4-chlorophenyl trifluoro borate 1-, potassium ion 4-chlorophenyl trifluoroboranuide
[B-](C1=CC=C(C=C1)Cl)(F)(F)F.[K+]
218.452
218.45
293°C to 295°C
661465-44-7
MFCD04115770
GXBSKMLVYJPMRT-UHFFFAOYSA-N
potassium;(4-chlorophenyl)-trifluoroboranuide
23682053
96%

Chemical Identifiers

661465-44-7
218.452
GXBSKMLVYJPMRT-UHFFFAOYSA-N
23682053
[B-](C1=CC=C(C=C1)Cl)(F)(F)F.[K+]
C6H4BClF3K
MFCD04115770
potassium 4-chlorophenyltrifluoroborate, potassium 4-chlorophenyl trifluoroborate, potassium 4-chlorophenyl trifluoroboranuide, pubchem11348, potassium 4-chlorophenyltrifluoroborate, potassium 4-chlorophenyl trifluoro borate 1-, potassium ion 4-chlorophenyl trifluoroboranuide
potassium;(4-chlorophenyl)-trifluoroboranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents