Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-benzyloxyphenyltrifluoroborate, 95%

$74.28 - $290.81

Chemical Identifiers

Chemical Name or Material Potassium 4-benzyloxyphenyltrifluoroborate
CAS 850623-47-1
Molecular Formula C13H11BF3KO
Formula Weight 290.13
MDL Number MFCD04115746
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5189406

  SDS
alfa aesar™
H5189406
5g Each for $290.81

AAH5189403

  SDS
alfa aesar™
H5189403
1g Each for $74.28
Description & Specifications

Specifications

Formula Weight 290.13
Chemical Name or Material Potassium 4-benzyloxyphenyltrifluoroborate
Assay 95%
Molecular Formula C13H11BF3KO
MDL Number MFCD04115746
CAS 850623-47-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.