missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-acetylphenyltrifluoroborate, 97%

$53.99 - $875.92

Chemical Identifiers

CAS 252726-24-2
Molecular Formula C8H7BF3KO
Molecular Weight (g/mol) 226.047
MDL Number MFCD07784376
InChI Key JRUXIRSHKHCTTK-UHFFFAOYSA-N
Synonym potassium 4-acetylphenyl trifluoro borate 1-
PubChem CID 23665673
IUPAC Name potassium;(4-acetylphenyl)-trifluoroboranuide
SMILES [B-](C1=CC=C(C=C1)C(=O)C)(F)(F)F.[K+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5584503 View Documents Alfa Aesar™
H5584503
1g Each for $53.99
 
AAH5584506 View Documents Alfa Aesar™
H5584506
5g Each for $215.36
 
AAH5584514 View Documents Alfa Aesar™
H5584514
25g Each for $875.92
 
Specifications

Chemical Identifiers

252726-24-2
226.047
JRUXIRSHKHCTTK-UHFFFAOYSA-N
23665673
[B-](C1=CC=C(C=C1)C(=O)C)(F)(F)F.[K+]
C8H7BF3KO
MFCD07784376
potassium 4-acetylphenyl trifluoro borate 1-
potassium;(4-acetylphenyl)-trifluoroboranuide

Specifications

252726-24-2
MFCD07784376
JRUXIRSHKHCTTK-UHFFFAOYSA-N
potassium;(4-acetylphenyl)-trifluoroboranuide
23665673
Powder
White
1g
C8H7BF3KO
potassium 4-acetylphenyl trifluoro borate 1-
[B-](C1=CC=C(C=C1)C(=O)C)(F)(F)F.[K+]
226.047
226.05
97%
290°C (decomposition)
Potassium 4-acetylphenyltrifluoroborate
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures