missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-acetylphenyltrifluoroborate, 97%

Manufacturer:  Alfa Aesar™ H5584514

 View more versions of this product

Catalog No. AAH5584514



Specifications

Specifications

Potassium 4-acetylphenyltrifluoroborate
C8H7BF3KO
potassium 4-acetylphenyl trifluoro borate 1-
[B-](C1=CC=C(C=C1)C(=O)C)(F)(F)F.[K+]
226.047
226.05
97%
290°C (decomposition)
252726-24-2
MFCD07784376
JRUXIRSHKHCTTK-UHFFFAOYSA-N
potassium;(4-acetylphenyl)-trifluoroboranuide
23665673
Powder
White
25g

Chemical Identifiers

252726-24-2
226.047
JRUXIRSHKHCTTK-UHFFFAOYSA-N
23665673
[B-](C1=CC=C(C=C1)C(=O)C)(F)(F)F.[K+]
C8H7BF3KO
MFCD07784376
potassium 4-acetylphenyl trifluoro borate 1-
potassium;(4-acetylphenyl)-trifluoroboranuide
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents