Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-(1-piperidinylmethyl)phenyltrifluoroborate, 95%

$90.55 - $90.55

Chemical Identifiers

Chemical Name or Material Potassium 4-(1-piperidinylmethyl)phenyltrifluoroborate
Molecular Formula C12H16BF3KN
Formula Weight 281.17
MDL Number MFCD16293905
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5189506

  SDS
alfa aesar™
H5189506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5189503

  SDS
alfa aesar™
H5189503
1g Each for $90.55
Description & Specifications

Specifications

Formula Weight 281.17
Health Hazard 1 Warning
Molecular Formula C12H16BF3KN
Chemical Name or Material Potassium 4-(1-piperidinylmethyl)phenyltrifluoroborate
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Assay 95%
MDL Number MFCD16293905
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.