Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-(1-piperidinylcarbonyl)phenyltrifluoroborate, 97%

$60.34 - $232.43

Chemical Identifiers

Chemical Name or Material Potassium 4-(1-piperidinylcarbonyl)phenyltrifluoroborate
CAS 1359865-98-7
Molecular Formula C12H14BF3KNO
Formula Weight 295.15
MDL Number MFCD11977715
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5114603

 
alfa aesar™
H5114603
1g Each for $60.34

AAH5114606

 
alfa aesar™
H5114606
5g Each for $232.43
Description & Specifications

Specifications

Formula Weight 295.15
Chemical Name or Material Potassium 4-(1-piperidinylcarbonyl)phenyltrifluoroborate
Assay 97%
Molecular Formula C12H14BF3KNO
MDL Number MFCD11977715
CAS 1359865-98-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.