missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 3-methoxyphenyltrifluoroborate, 98%

Manufacturer:  Alfa Aesar™ H5320003

 View more versions of this product

Catalog No. AAH5320003



Specifications

Specifications

Potassium 3-methoxyphenyltrifluoroborate
1g
C7H7BF3KO
potassium 3-methoxyphenyl trifluoroborate, potassium trifluoro 3-methoxyphenyl borate, potassium 3-methoxyphenyltrifluoroborate, potassium 3-methoxypheny trifluoroborate, potassium trifluoro 3-methoxyphenyl boranuide, amtb102, trifluoro 3-methoxyphenyl potassioboron v, potassium trifluoro-3-methoxyphenyl boranuide, potassium trifluoro 3-methoxyphenyl borate 1-, potassium ion trifluoro 3-methoxyphenyl boranuide
[B-](C1=CC(=CC=C1)OC)(F)(F)F.[K+]
214.036
214.04
164°C to 168°C
438553-44-7
MFCD04039954
GKDCWEPJOSDGHY-UHFFFAOYSA-N
potassium;trifluoro-(3-methoxyphenyl)boranuide
23697282
98%

Chemical Identifiers

438553-44-7
214.036
GKDCWEPJOSDGHY-UHFFFAOYSA-N
23697282
[B-](C1=CC(=CC=C1)OC)(F)(F)F.[K+]
C7H7BF3KO
MFCD04039954
potassium 3-methoxyphenyl trifluoroborate, potassium trifluoro 3-methoxyphenyl borate, potassium 3-methoxyphenyltrifluoroborate, potassium 3-methoxypheny trifluoroborate, potassium trifluoro 3-methoxyphenyl boranuide, amtb102, trifluoro 3-methoxyphenyl potassioboron v, potassium trifluoro-3-methoxyphenyl boranuide, potassium trifluoro 3-methoxyphenyl borate 1-, potassium ion trifluoro 3-methoxyphenyl boranuide
potassium;trifluoro-(3-methoxyphenyl)boranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents