Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 3-formylphenyltrifluoroborate, 97%

$44.01 - $135.46

Chemical Identifiers

Chemical Name or Material Potassium 3-formylphenyltrifluoroborate
CAS 871231-44-6
Molecular Formula C7H5BF3KO
Formula Weight 212.02
MDL Number MFCD07784378
Synonym 3-Formylbenzenetrifluoroboric acid potassium salt; 3-Formylphenyltrifluoroboric acid potassium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2812503

 
alfa aesar™
H2812503
1g Each for $44.01

AAH2812506

 
alfa aesar™
H2812506
5g Each for $135.46
Description & Specifications

Specifications

Formula Weight 212.02
Chemical Name or Material Potassium 3-formylphenyltrifluoroborate
Assay 97%
Molecular Formula C7H5BF3KO
MDL Number MFCD07784378
CAS 871231-44-6
Synonym 3-Formylbenzenetrifluoroboric acid potassium salt; 3-Formylphenyltrifluoroboric acid potassium salt
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈200°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.