missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 3-chlorophenyltrifluoroborate, 96%

Manufacturer:  Alfa Aesar™ H5314403

 View more versions of this product

Catalog No. AAH5314403



Specifications

Specifications

Potassium 3-chlorophenyltrifluoroborate
1g
C6H4BClF3K
potassium 3-chlorophenyl trifluoroborate, potassium 3-chlorophenyl trifluoroboranuide, potassium 3-chlorophenyltrifluoroborate, amtb707, potassium 3-chlorophenyl-trifluoroboranuide, potassium 3-chlorophenyl trifluoro borate 1-, potassium ion 3-chlorophenyl trifluoroboranuide, potassium 3-chlorophenyl-tris fluoranyl boranuide
[B-](C1=CC(=CC=C1)Cl)(F)(F)F.[K+]
218.452
218.45
76°C to 80°C
411206-75-2
MFCD04115754
LRJCVRYGKNDLAD-UHFFFAOYSA-N
potassium;(3-chlorophenyl)-trifluoroboranuide
23677818
96%

Chemical Identifiers

411206-75-2
218.452
LRJCVRYGKNDLAD-UHFFFAOYSA-N
23677818
[B-](C1=CC(=CC=C1)Cl)(F)(F)F.[K+]
C6H4BClF3K
MFCD04115754
potassium 3-chlorophenyl trifluoroborate, potassium 3-chlorophenyl trifluoroboranuide, potassium 3-chlorophenyltrifluoroborate, amtb707, potassium 3-chlorophenyl-trifluoroboranuide, potassium 3-chlorophenyl trifluoro borate 1-, potassium ion 3-chlorophenyl trifluoroboranuide, potassium 3-chlorophenyl-tris fluoranyl boranuide
potassium;(3-chlorophenyl)-trifluoroboranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents