Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 3-carboxyphenyltrifluoroborate, 95%

$60.72 - $226.01

Chemical Identifiers

Chemical Name or Material Potassium 3-carboxyphenyltrifluoroborate
CAS 850313-91-6
Molecular Formula C7H5BF3KO2
Formula Weight 228.02
MDL Number MFCD07784423
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5188503

  SDS
alfa aesar™
H5188503
1g Each for $60.72

AAH5188506

  SDS
alfa aesar™
H5188506
5g Each for $226.01
Description & Specifications

Specifications

Formula Weight 228.02
Chemical Name or Material Potassium 3-carboxyphenyltrifluoroborate
Assay 95%
Molecular Formula C7H5BF3KO2
MDL Number MFCD07784423
CAS 850313-91-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.