missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 3-bromophenyltrifluoroborate, 97%

$66.46 - $245.20

Chemical Identifiers

CAS 374564-34-8
Molecular Formula C6H4BBrF3K
Molecular Weight (g/mol) 262.906
MDL Number MFCD02093977
InChI Key NIDYRVJZTADCGO-UHFFFAOYSA-N
Synonym potassium 3-bromophenyl trifluoroborate, potassium 3-bromophenyltrifluoroborate, potassium 3-bromophenyl trifluoroboranuide, amtb081, potassium 3-bromophenyl-trifluoroboranuide, potassium ion 3-bromophenyl trifluoroboranuide, potassium 3-bromophenyl trifluoro borate 1-, potassium 3-bromophenyl-tris fluoranyl boranuide
PubChem CID 23677642
IUPAC Name potassium;(3-bromophenyl)-trifluoroboranuide
SMILES [B-](C1=CC(=CC=C1)Br)(F)(F)F.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1796603 View Documents Alfa Aesar™
L1796603
1g Each for $66.46
 
AAL1796606 View Documents Alfa Aesar™
L1796606
5g Each for $245.20
 
Specifications

Chemical Identifiers

374564-34-8
262.906
NIDYRVJZTADCGO-UHFFFAOYSA-N
23677642
[B-](C1=CC(=CC=C1)Br)(F)(F)F.[K+]
C6H4BBrF3K
MFCD02093977
potassium 3-bromophenyl trifluoroborate, potassium 3-bromophenyltrifluoroborate, potassium 3-bromophenyl trifluoroboranuide, amtb081, potassium 3-bromophenyl-trifluoroboranuide, potassium ion 3-bromophenyl trifluoroboranuide, potassium 3-bromophenyl trifluoro borate 1-, potassium 3-bromophenyl-tris fluoranyl boranuide
potassium;(3-bromophenyl)-trifluoroboranuide

Specifications

178°C to 182°C
374564-34-8
MFCD02093977
NIDYRVJZTADCGO-UHFFFAOYSA-N
potassium;(3-bromophenyl)-trifluoroboranuide
23677642
97%
1g
C6H4BBrF3K
potassium 3-bromophenyl trifluoroborate, potassium 3-bromophenyltrifluoroborate, potassium 3-bromophenyl trifluoroboranuide, amtb081, potassium 3-bromophenyl-trifluoroboranuide, potassium ion 3-bromophenyl trifluoroboranuide, potassium 3-bromophenyl trifluoro borate 1-, potassium 3-bromophenyl-tris fluoranyl boranuide
[B-](C1=CC(=CC=C1)Br)(F)(F)F.[K+]
262.906
262.91
Potassium 3-bromophenyltrifluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning