missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium 3,5-bis(trifluoromethyl)phenytrifluoroborate, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5311306
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsThos chemical has been used for inserting a 3,5-bis(trifluoromethyl)phenyl group into the position of a halogen or triflate group of various arenes and various heterocycles in Pd-catalyzed Suzuli-Miyaura cross-coupling reactions.
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications
Potassium 3,5-bis(trifluoromethyl)phenytrifluoroborate | |
>300°C | |
MFCD03701625 | |
potassium 3,5-bis trifluoromethyl phenyltrifluoroborate, potassium 3,5-bis trifluoromethyl phenyl trifluoroborate, potassium 3,5-bis trifluoromethyl phenyl trifluoroboranuide, pubchem11421, c8h3bcf9k, amtb724, potassium 3,5-bis trifluoromethyl phenytrifluoroborate, potassium 3,5-bistrifluoromethylphenyltrifluoroborate, potassium 3,5-bis trifluoromethyl phenyl-trifluoroboranuide | |
[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(F)(F)F.[K+] | |
320.006 | |
320.01 |
166328-09-2 | |
C8H3BF9K | |
5 g | |
WCHMTVMZBOZGCS-UHFFFAOYSA-N | |
potassium;[3,5-bis(trifluoromethyl)phenyl]-trifluoroboranuide | |
23668091 | |
96% |
Chemical Identifiers
166328-09-2 | |
320.006 | |
WCHMTVMZBOZGCS-UHFFFAOYSA-N | |
23668091 | |
[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(F)(F)F.[K+] |
C8H3BF9K | |
MFCD03701625 | |
potassium 3,5-bis trifluoromethyl phenyltrifluoroborate, potassium 3,5-bis trifluoromethyl phenyl trifluoroborate, potassium 3,5-bis trifluoromethyl phenyl trifluoroboranuide, pubchem11421, c8h3bcf9k, amtb724, potassium 3,5-bis trifluoromethyl phenytrifluoroborate, potassium 3,5-bistrifluoromethylphenyltrifluoroborate, potassium 3,5-bis trifluoromethyl phenyl-trifluoroboranuide | |
potassium;[3,5-bis(trifluoromethyl)phenyl]-trifluoroboranuide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures