missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 3,5-bis(trifluoromethyl)phenytrifluoroborate, 96%

Manufacturer:  Alfa Aesar™ H5311303

 View more versions of this product

Catalog No. AAH5311303



Description

Description

Applications
Thos chemical has been used for inserting a 3,5-bis(trifluoromethyl)phenyl group into the position of a halogen or triflate group of various arenes and various heterocycles in Pd-catalyzed Suzuli-Miyaura cross-coupling reactions.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Specifications

Potassium 3,5-bis(trifluoromethyl)phenytrifluoroborate
1g
C8H3BF9K
potassium 3,5-bis trifluoromethyl phenyltrifluoroborate, potassium 3,5-bis trifluoromethyl phenyl trifluoroborate, potassium 3,5-bis trifluoromethyl phenyl trifluoroborate, potassium 3,5-bis trifluoromethyl phenyl trifluoroboranuide, pubchem11421, c8h3bcf9k, amtb724, potassium 3,5-bis trifluoromethyl phenytrifluoroborate, potassium 3,5-bistrifluoromethylphenyltrifluoroborate, potassium 3,5-bis trifluoromethyl phenyl-trifluoroboranuide
[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(F)(F)F.[K+]
320.006
320.01
>300°C
166328-09-2
MFCD03701625
WCHMTVMZBOZGCS-UHFFFAOYSA-N
potassium;[3,5-bis(trifluoromethyl)phenyl]-trifluoroboranuide
23668091
96%

Chemical Identifiers

166328-09-2
320.006
WCHMTVMZBOZGCS-UHFFFAOYSA-N
23668091
[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(F)(F)F.[K+]
C8H3BF9K
MFCD03701625
potassium 3,5-bis trifluoromethyl phenyltrifluoroborate, potassium 3,5-bis trifluoromethyl phenyl trifluoroborate, potassium 3,5-bis trifluoromethyl phenyl trifluoroborate, potassium 3,5-bis trifluoromethyl phenyl trifluoroboranuide, pubchem11421, c8h3bcf9k, amtb724, potassium 3,5-bis trifluoromethyl phenytrifluoroborate, potassium 3,5-bistrifluoromethylphenyltrifluoroborate, potassium 3,5-bis trifluoromethyl phenyl-trifluoroboranuide
potassium;[3,5-bis(trifluoromethyl)phenyl]-trifluoroboranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents