missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 3,4-dichlorophenyltrifluoroborate, 96%

Manufacturer:  Alfa Aesar™ H5321903

 View more versions of this product

Catalog No. AAH5321903



Specifications

Specifications

Potassium 3,4-dichlorophenyltrifluoroborate
1g
C6H3BCl2F3K
potassium 3,4-dichlorophenyl trifluoroborate, potassium 3,4-dichlorophenyl trifluoroboranuide, potassium 3,4-dichlorophenyltrifluoroborate, c6h3bcl2f3.k, potassium 3,4-dichlorophenyl trifluoroborate, potassium 3,4-dichlorophenyl-trifluoroboranuide, potassium ion 3,4-dichlorophenyl trifluoroboranuide, potassium 3,4-dichlorophenyl trifluoro borate 1-, potassium 3,4-dichlorophenyl-tris fluoranyl boranuide
[B-](C1=CC(=C(C=C1)Cl)Cl)(F)(F)F.[K+]
252.894
252.9
178°C to 180°C
850623-68-6
MFCD04115763
RTZOKQYJVLRZFW-UHFFFAOYSA-N
potassium;(3,4-dichlorophenyl)-trifluoroboranuide
44717229
96%

Chemical Identifiers

850623-68-6
252.894
RTZOKQYJVLRZFW-UHFFFAOYSA-N
44717229
[B-](C1=CC(=C(C=C1)Cl)Cl)(F)(F)F.[K+]
C6H3BCl2F3K
MFCD04115763
potassium 3,4-dichlorophenyl trifluoroborate, potassium 3,4-dichlorophenyl trifluoroboranuide, potassium 3,4-dichlorophenyltrifluoroborate, c6h3bcl2f3.k, potassium 3,4-dichlorophenyl trifluoroborate, potassium 3,4-dichlorophenyl-trifluoroboranuide, potassium ion 3,4-dichlorophenyl trifluoroboranuide, potassium 3,4-dichlorophenyl trifluoro borate 1-, potassium 3,4-dichlorophenyl-tris fluoranyl boranuide
potassium;(3,4-dichlorophenyl)-trifluoroboranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents