Please login to your online account to display your discounted pricing

Potassium (2-nitrophenyl)trifluoroborate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Potassium (2-nitrophenyl)trifluoroborate
Assay Percent Range 97%
Molecular Formula C6H4BF3KNO2
Formula Weight 229.01
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444850050

 
thermo fisher scientific
AC444850050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444850010

 
thermo fisher scientific
AC444850010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.01
Quantity 5g
Chemical Name or Material Potassium (2-nitrophenyl)trifluoroborate
Assay Percent Range 97%
Molecular Formula C6H4BF3KNO2
CAS 850623-64-2