missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium (2-nitrophenyl)trifluoroborate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444850010
Specifications
Potassium (2-nitrophenyl)trifluoroborate | |
226°C to 228°C | |
850623-64-2 | |
C6H4BF3KNO2 | |
DZHYOFKWMHZKBK-UHFFFAOYSA-N | |
potassium;trifluoro-(2-nitrophenyl)boranuide | |
2782854 | |
97% |
Glass Bottle | |
1g | |
97% | |
amtb936, potassium 2-nitrophenyl trifluoroborate, potassium 2-nitrophenyltrifluoroborate, potassium ion trifluoro 2-nitrophenyl boranuide, potassium trifluoro 2-nitrophenyl boranuide, potassium trifluoro 2-nitrophenyl borate, potassium trifluoro-2-nitrophenyl boranuide, potassium tris fluoranyl-2-nitrophenyl boranuide | |
[B-](C1=CC=CC=C1[N+](=O)[O-])(F)(F)F.[K+] | |
229.01 | |
229.01 |
Chemical Identifiers
850623-64-2 | |
229.01 | |
amtb936, potassium 2-nitrophenyl trifluoroborate, potassium 2-nitrophenyltrifluoroborate, potassium ion trifluoro 2-nitrophenyl boranuide, potassium trifluoro 2-nitrophenyl boranuide, potassium trifluoro 2-nitrophenyl borate, potassium trifluoro-2-nitrophenyl boranuide, potassium tris fluoranyl-2-nitrophenyl boranuide | |
potassium;trifluoro-(2-nitrophenyl)boranuide |
C6H4BF3KNO2 | |
DZHYOFKWMHZKBK-UHFFFAOYSA-N | |
2782854 | |
[B-](C1=CC=CC=C1[N+](=O)[O-])(F)(F)F.[K+] |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning