missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 2-methylphenyltrifluoroborate, 96%

Manufacturer:  Alfa Aesar™ H5321406

 View more versions of this product

Catalog No. AAH5321406



Specifications

Specifications

Potassium 2-methylphenyltrifluoroborate
5g
C7H7BF3K
potassium trifluoro o-tolyl borate, potassium 2-methylphenyl trifluoroborate, potassium o-tolyltrifluoroborate, potassium trifluoro 2-methylphenyl boranuide, potassium 2-methylphenyltrifluoroborate, pubchem11432, potassiumo-tolyltrifluoroborate, trifluoro 2-methylphenyl potassioboron v, potassium trifluoro-2-methylphenyl boranuide, potassium trifluoro 2-methylphenyl borate 1-
[B-](C1=CC=CC=C1C)(F)(F)F.[K+]
198.037
198.04
90°C to 94°C
274257-34-0
MFCD03790006
ROMUTKWSUQKVLS-UHFFFAOYSA-N
potassium;trifluoro-(2-methylphenyl)boranuide
23674703
96%

Chemical Identifiers

274257-34-0
198.037
ROMUTKWSUQKVLS-UHFFFAOYSA-N
23674703
[B-](C1=CC=CC=C1C)(F)(F)F.[K+]
C7H7BF3K
MFCD03790006
potassium trifluoro o-tolyl borate, potassium 2-methylphenyl trifluoroborate, potassium o-tolyltrifluoroborate, potassium trifluoro 2-methylphenyl boranuide, potassium 2-methylphenyltrifluoroborate, pubchem11432, potassiumo-tolyltrifluoroborate, trifluoro 2-methylphenyl potassioboron v, potassium trifluoro-2-methylphenyl boranuide, potassium trifluoro 2-methylphenyl borate 1-
potassium;trifluoro-(2-methylphenyl)boranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents