Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 2-formylphenyltrifluoroborate, 97%

$115.27 - $115.27

Chemical Identifiers

Chemical Name or Material Potassium 2-formylphenyltrifluoroborate
CAS 192863-39-1
Molecular Formula C7H5BF3KO
Formula Weight 212.02
MDL Number MFCD02093979
Beilstein 7783590
Synonym 2-Formylbenzenetrifluoroboric acid potassium salt; 2-Formylphenyltrifluoroboric acid potassium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1796803

  SDS
alfa aesar™
L1796803
1g Each for $115.27
Description & Specifications

Specifications

Formula Weight 212.02
Chemical Name or Material Potassium 2-formylphenyltrifluoroborate
CAS 192863-39-1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Assay 97%
MDL Number MFCD02093979
Molecular Formula C7H5BF3KO
Beilstein 7783590
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Synonym 2-Formylbenzenetrifluoroboric acid potassium salt; 2-Formylphenyltrifluoroboric acid potassium salt
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.