missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium 2-Ethylhexanoate, TCI America™

Manufacturer:  TCI America E012125G

 View more versions of this product

Catalog No. E012125G



Specifications

Specifications

Potassium 2-Ethylhexanoate
25g
C8H15KO2
potassium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, potassium salt, 2-ethylhexanoic acid potassium salt, 2-ethylhexanoic acid, potassium salt, hexanoic acid, 2-ethyl-, potassium salt 1:1, potassium2-ethylhexanoate, dsstox_cid_7525, dsstox_rid_78488, dsstox_gsid_27525, ksc493c3p
CCCCC(CC)C(=O)[O-].[K+]
182.304
182.30
≥95.0% (T)
White
3164-85-0
MFCD00045896
ZUFQCVZBBNZMKD-UHFFFAOYSA-M
potassium;2-ethylhexanoate
23669619
Crystal-Lump at 20°C

Chemical Identifiers

3164-85-0
182.304
ZUFQCVZBBNZMKD-UHFFFAOYSA-M
23669619
CCCCC(CC)C(=O)[O-].[K+]
C8H15KO2
MFCD00045896
potassium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, potassium salt, 2-ethylhexanoic acid potassium salt, 2-ethylhexanoic acid, potassium salt, hexanoic acid, 2-ethyl-, potassium salt 1:1, potassium2-ethylhexanoate, dsstox_cid_7525, dsstox_rid_78488, dsstox_gsid_27525, ksc493c3p
potassium;2-ethylhexanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0611

TSCA : Yes

Documents