missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium 2-ethylhexanoate hydrate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03944006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium 2-ethylhexanoate hydrate | |
Odorless | |
K[OOCCH(C2H5)C4H9]·xH2O | |
5 g | |
potassium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, potassium salt, 2-ethylhexanoic acid potassium salt, 2-ethylhexanoic acid, potassium salt, hexanoic acid, 2-ethyl-, potassium salt 1:1, potassium2-ethylhexanoate, dsstox_cid_7525, dsstox_rid_78488, dsstox_gsid_27525, ksc493c3p | |
[K+].CCCCC(CC)C([O-])=O | |
182.30 | |
182.30 (Anhydrous) | |
Lump or Semiliquid |
3164-85-0 | |
C8H15KO2 | |
MFCD00045896 | |
Moisture sensitive | |
ZUFQCVZBBNZMKD-UHFFFAOYNA-M | |
potassium;2-ethylhexanoate | |
23669619 | |
95% |
Chemical Identifiers
3164-85-0 | |
182.30 | |
ZUFQCVZBBNZMKD-UHFFFAOYNA-M | |
[K+].CCCCC(CC)C([O-])=O |
C8H15KO2 | |
MFCD00045896 | |
23669619 |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P264b-P281-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P501c
H315-H318-H361
EINECSNumber : 221-625-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only