Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 2,4-pentanedionate hydrate, 97%

$60.88 - $711.19

Chemical Identifiers

Chemical Name or Material Potassium 2,4-pentanedionate hydrate
CAS 57402-46-7
Molecular Formula C5H7KO2·xH2O
Formula Weight 138.21 (Anhydrous)
MDL Number MFCD00150167
Synonym Potassium acetylacetonate hemihydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2416514

  SDS
alfa aesar™
B2416514
25g Each for $60.88

AAB2416536

  SDS
alfa aesar™
B2416536
500g Each for $711.19

AAB2416522

  SDS
alfa aesar™
B2416522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Potassium 2,4-pentanedionate hydrate
CAS 57402-46-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C5H7KO2·xH2O
Formula Weight 138.21 (Anhydrous)
MDL Number MFCD00150167
Synonym Potassium acetylacetonate hemihydrate
Melting Point ≈232°C (dec.)
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.