Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 2,4-bis(trifluoromethyl)phenyltrifluoroborate, 95%

Chemical Identifiers

Chemical Name or Material Potassium 2,4-bis(trifluoromethyl)phenyltrifluoroborate
CAS 1150655-10-9
Formula Weight 320.01
MDL Number MFCD07368252
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6324603

  SDS
alfa aesar™
H6324603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6324606

  SDS
alfa aesar™
H6324606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 320.01
Melting Point 173° to 178°C
Health Hazard 1 Warning
Assay 95%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Storage Note 1 2° to 8°C
MDL Number MFCD07368252
Quantity 1g
Chemical Name or Material Potassium 2,4-bis(trifluoromethyl)phenyltrifluoroborate
Physical Form Solid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1150655-10-9
Color White to pale brown
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C