missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 2,4-bis(trifluoromethyl)phenyltrifluoroborate, 95%

Manufacturer:  Alfa Aesar™ H6324606

 View more versions of this product

Catalog No. AAH6324606


 

Specifications

Specifications

Potassium 2,4-bis(trifluoromethyl)phenyltrifluoroborate
95%
MFCD07368252
[B-](C1=C(C=C(C=C1)C(F)(F)F)C(F)(F)F)(F)(F)F.[K+]
320.006
320.01
Brown to White
5g
1150655-10-9
C8H3BF9K
OXVQSBLFWHNZJA-UHFFFAOYSA-N
potassium;[2,4-bis(trifluoromethyl)phenyl]-trifluoroboranuide
44717658
Solid
173°C to 178°C

Chemical Identifiers

1150655-10-9
320.006
OXVQSBLFWHNZJA-UHFFFAOYSA-N
C8H3BF9K
MFCD07368252
44717658
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents