Please login to your online account to display your discounted pricing

Potassium, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$368.56 - $425.91

Chemical Identifiers

Chemical Name or Material Potassium, 1000μg/mL (0.10% w/v)
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula K
Formula Weight 39.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003700

 
avantor performance
577404
Poly Bottle 100mL Each for $425.91

02003701

 
avantor performance
646404
Narrow Mouth Poly Bottle 150mL Each for $368.56
Description & Specifications

Specifications

Formula Weight 39.1
Chemical Name or Material Potassium, 1000μg/mL (0.10% w/v)
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula K
Packaging Poly Bottle
Quantity 100mL
Name Note Plasma Standard
Specific Gravity 1L = 1.00kg