missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PortaFab CleanLine™ S3000 Modular Wall System Cleanroom - ISO 6

Outfit existing facilities or create freestanding envelope structures with a cleanroom or environmental enclosure

Specifications

Certifications/Compliance ISO 6 cleanroom
Color White
Housing Material 24G Sheet Steel, Polyester Baked Enamel
View More Specs

Disclaimers:

Local municipalities, townships and villages independently establish, adopt and interpret building codes; Therefore, PortaFab cannot represent that this structure will meet all local building codes; It is your responsibility to determine code compliance.

Requires: 122cm (48 in.) T8 bulbs: four per light fixture (not included)



Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W x H) Filter Lights Outlets Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Filter Lights Outlets Price Quantity  
19156502 PortaFab
6CR816
96 x 192 x 120 in. (240 x 490 x 300cm) 5 HEPA 2 4 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156504 PortaFab
6CR824
96 x 288 x 120 in. (240 x 730 x 300cm) 8 HEPA 3 6 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156501 PortaFab
6CR812
96 x 144 x 120 in. (240 x 370 x 300cm) 4 HEPA 1 3 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156503 PortaFab
6CR820
96 x 240 x 120 in. (240 x 610 x 300cm) 6 HEPA 2 4 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156505 PortaFab
6CR1212
144 x 144 x 120 in. (370 x 370 x 300cm) 6 HEPA 2 4 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156508 PortaFab
6CR1224
144 x 288 x 120 in. (370 x 730 x 300cm) 11 HEPA 4 6 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156506 PortaFab
6CR1216
144 x 192 x 120 in. (370 x 490 x 300cm) 8 HEPA 3 4 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156510 PortaFab
6CR1620
192 x 240 x 120 in. (490 x 610 x 300cm) 13 HEPA 5 6 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156507 PortaFab
6CR1220
144 x 240 x 120 in. (370 x 610 x 300cm) 9 HEPA 4 6 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156513 PortaFab
6CR1636
192 x 432 x 120 in. (490 x 1100 x 300cm) 23 HEPA 8 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156509 PortaFab
6CR1616
192 x 192 x 120 in. (490 x 490 x 300cm) 10 HEPA 4 6 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156512 PortaFab
6CR1630
192 x 360 x 120 in. (490 x 914 x 300cm) 19 HEPA 7 6 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156511 PortaFab
6CR1624
192 x 288 x 120 in. (490 x 730 x 300cm) 15 HEPA 6 6 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156514 PortaFab
6CR2020
240 x 240 x 120 in. (610 x 610 x 300cm) 16 HEPA 7 6 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156516 PortaFab
6CR2030
240 x 360 x 120 in. (610 x 914 x 300cm) 23 HEPA 8 8 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156517 PortaFab
6CR2036
240 x 432 x 120 in. (610 x 1100 x 300cm) 28 HEPA 10 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156515 PortaFab
6CR2024
240 x 288 x 120 in. (610 x 730 x 300cm) 19 HEPA 8 6 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156521 PortaFab
6CR2436
288 x 432 x 120 in. (730 x 1100 x 300cm) 34 HEPA 12 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156520 PortaFab
6CR2430
288 x 360 x 120 in. (730 x 914 x 300cm) 28 HEPA 10 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156519 PortaFab
6CR2424
288 x 288 x 120 in. (730 x 730 x 300cm) 23 HEPA 8 8 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156518 PortaFab
6CR2048
240 x 576 x 120 in. (610 x 1460 x 300cm) 38 HEPA 14 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156522 PortaFab
6CR2448
288 x 576 x 120 in. (730 x 1460 x 300cm) 45 HEPA 16 12 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156523 PortaFab
6CR2460
288 x 720 x 120 in. (730 x 1830 x 300cm) 56 HEPA 20 12 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156524 PortaFab
6CR3030
360 x 360 x 120 in. (910 x 914 x 300cm) 35 HEPA 12 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156529 PortaFab
6CR4030
480 x 360 x 120 in. (1220 x 914 x 300cm) 47 HEPA 16 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156528 PortaFab
6CR4024
480 x 288 x 120 in. (1220 x 730 x 300cm) 38 HEPA 14 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156527 PortaFab
6CR3060
360 x 720 x 120 in. (910 x 1830 x 300cm) 70 HEPA 24 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156525 PortaFab
6CR3036
360 x 432 x 120 in. (910 x 1100 x 300cm) 42 HEPA 16 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156530 PortaFab
6CR4036
480 x 432 x 120 in. (1220 x 1100 x 300cm) 56 HEPA 20 12 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156526 PortaFab
6CR3048
360 x 576 x 120 in. (910 x 1460 x 300cm) 56 HEPA 20 12 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156531 PortaFab
6CR4048
480 x 576 x 120 in. (1220 x 1460 x 300cm) 75 HEPA 26 12 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19156532 PortaFab
6CR4060
480 x 720 x 120 in. (1220 x 1830 x 300cm) 94 HEPA 32 14 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Non-progressive construction allows easy panel change or structure relocation
  • 7.6cm (3 in.) thick walls are constructed of 24 ga sheet steel that is painted with polyester baked enamel on each side; steel is laminated to 1.27cm (0.5 in.) gypsum with a 4.9cm (1.94 in.) polystyrene core
  • Studs are 6063-T5 aluminum extrusions with a 201R1 anodized clear finish
  • Depending on size, some units may require interior columns
  • Color is white
  • Buildings are 3m (10 ft.) high; rooms do not come with a floor
  • Corrugated steel load-bearing roof holds mechanical equipment: 25psf load
  • Roof support: structural steel I-beams located on 8 ft. center
  • Acoustical ceiling: 61 x 192cm (24 x 48 in.) Class A noncombustible acoustical ceiling with 3.8cm (1.5 in.) gasketed grid and vinyl faced ceiling tiles
Entry
  • Seamless single steel door on a steel frame
  • W x H x D: .9 x 2.1 x .04m (36 x 84 x 1.75 in.)
  • Door has a 61 x 61 x .6cm (24 x 24 x 0.25 in.) thick tempered glass window, lever lockset, hinges, sweep and door closer
Electricals/Ventilation
  • Quantities of fittings vary based on room size, see ordering table below for more information
  • Modular wired electrical package includes: wall switch, 61 x 122cm (24 x 48 in.) recessed fluorescent light fixture(s) with acrylic lens cover, 110V outlet(s), 61 x 122cm (24 x 48 in.) 115V fan filter unit(s), handy boxes and circuit breaker panel(s) (with breakers)
  • Wiring studs permit wiring access on one side
  • HEPA filter(s)
  • 46 x 61cm (18 x 24 in.) and/or 61 x 61cm (24 x 24 in.) return air grill(s)

Applications:

Single pass and recirculating cleanrooms ; Cost effective enough for simpler applications, such as basic process segmentation or controlling air quality, temperature or humidity

Compliance

Meets the specifications of ISO 6 cleanrooms.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

ISO 6 cleanroom
White
24G Sheet Steel, Polyester Baked Enamel