Please login to your online account to display your discounted pricing

Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  418140050

 View more versions of this product

Catalog No. AC418140050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥21.2% (K)
Physical Form Crystalline Powder
Color Off-white to beige-brownish
Quantity 5g
Chemical Name or Material Polyvinylsulfuric acid potassium salt
Linear Formula (-CH2CH(OSO3K)-)n
CAS 26837-42-3
Synonym Polyvinyl sulfate potassium salt
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material Polyvinylsulfuric acid potassium salt
Assay ≥21.2% (K)
Linear Formula (-CH2CH(OSO3K)-)n
Synonym Polyvinyl sulfate potassium salt