Please login to your online account to display your discounted pricing

Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC418141000

 View more versions of this product

Catalog No. AC418141000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥21.2% (K)
Color Off-white to beige-brownish
Quantity 100g
Synonym Polyvinyl Sulfate Potassium Salt
Chemical Name or Material Polyvinylsulfuric acid potassium salt
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 26837-42-3
Linear Formula (-CH2CH(OSO3K)-)n

Specifications

Chemical Name or Material Polyvinylsulfuric acid potassium salt
Assay ≥21.2% (K)
Linear Formula (-CH2CH(OSO3K)-)n
Synonym Polyvinyl Sulfate Potassium Salt