Learn More
Polyvinylpyrrolidone, M.W. 10,000, Thermo Scientific Chemicals
A cryoprotectant, also useful in plant cell culture research
Supplier: Thermo Scientific Chemicals J6038236
Description
Polyvinylpyrrolidone is a polymer used as a pharmaceutical aid, complexing agent, and solubilizer. PVP was used as a plasma volume expander for trauma victims. It acts as diluent, solvent and cleaning agent in various kinds of industries. It also acts as an adjuvant, emulsifier, filtering aid, tableting aid, thickener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
Powder | |
(C6H9NO)n | |
14,7697 | |
Soluble in water and most organic solvents. | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
1-ethenylpyrrolidin-2-one | |
6917 | |
10 kd |
Polyvinylpyrrolidone | |
9003-39-8 | |
MFCD01076626 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
500 g | |
*-CC(-*)N1CCCC1=O | |
111.14 | |
CHEBI:82551 |
Chemical Identifiers
9003-39-8 | |
111.14 | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
6917 | |
1-ethenylpyrrolidin-2-one |
(C6H9NO)n | |
MFCD01076626 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
CHEBI:82551 | |
*-CC(-*)N1CCCC1=O |
Safety and Handling
H500
RTECSNumber : TR8370000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only