missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyvinylpyrrolidone-iodine complex

Manufacturer:  Alfa Aesar™ 4578230

 View more versions of this product

Catalog No. AA4578230



Specifications

Specifications

Polyvinylpyrrolidone-iodine complex
C6H9I2NO
14,7698
CPKVUHPKYQGHMW-UHFFFAOYSA-N
1-ethenylpyrrolidin-2-one;molecular iodine
410087
Brown-Yellow
250g
25655-41-8
MFCD00084483
povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex
C=CN1CCCC1=O.II
364.953
Powder
300°C (decomposition)

Chemical Identifiers

25655-41-8
364.953
CPKVUHPKYQGHMW-UHFFFAOYSA-N
410087
C=CN1CCCC1=O.II
C6H9I2NO
MFCD00084483
povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex
1-ethenylpyrrolidin-2-one;molecular iodine
Safety and Handling

Safety and Handling

RTECSNumber : TR1579600

TSCA : TSCA

Documents