missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyvinylpyrrolidone-iodine complex, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04578218

 View more versions of this product

Catalog No. AA4578218



Description

Description

Polyvinylpyrrolidone-iodine complex is used as a slow iodine release formulation that iodinates lipids and oxidizes cell compounds such as tyrosine side chains. It is utilized to study microbial decontamination of human donor eyes as well as inactivation of infectious hematopoietic necrosis virus. It is also used as a carbon nanotube coating agent to enhance water solubility. It serves as a topical antiseptic in surgery, as an aerosol for skin and mucous membrane infections and is used to prevent infection in wounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Brown-Yellow
Powder
300°C (decomposition)
MFCD00084483
povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex
50 g
II.*-CC(-*)N1CCCC1=O
364.95
Polyvinylpyrrolidone-iodine complex
25655-41-8
(C6H9NO)nI2
14,7698
Soluble in water,ethyl alcohol,isopropyl alcohol,polyethylene glycol and glycerol. Insoluble in chloroform,carbon tetrachloride,hexane,acetone and petroleum ether.
CPKVUHPKYQGHMW-UHFFFAOYSA-N
1-ethenylpyrrolidin-2-one;molecular iodine
410087

Chemical Identifiers

25655-41-8
364.95
CPKVUHPKYQGHMW-UHFFFAOYSA-N
410087
(C6H9NO)nI2
MFCD00084483
povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex
II.*-CC(-*)N1CCCC1=O
Safety and Handling

Safety and Handling

RTECSNumber : TR1579600

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only