missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyvinylpyrrolidone, cross-linked, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 227480250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
5 to 8 (1% aq. soln.) | |
25249-54-1 | |
MFCD00149016 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
1-ethenylpyrrolidin-2-one | |
6917 | |
11 to 12.8% (nitrogen)(on substance as is) | |
5% max. | |
5% max. (K.F.) |
Polyvinylpyrrolidone | |
Micro-Crystalline Powder | |
(C6H9NO)n | |
14, 7697 | |
25 g | |
*-CC(-*)N1CCCC1=O | |
111.14 | |
CHEBI:82551 | |
Authentic | |
Plastic bottle |
Chemical Identifiers
25249-54-1 | |
111.14 | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
6917 | |
1-ethenylpyrrolidin-2-one |
(C6H9NO)n | |
MFCD00149016 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
CHEBI:82551 | |
*-CC(-*)N1CCCC1=O |
RUO – Research Use Only