missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyvinylpyrrolidone, average M.W. 3500, K12, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC276140010
Specifications
Polyvinylpyrrolidone | |
9003-39-8 | |
K-value: 10.2 to 13.8 | |
MFCD01076626 | |
B color: < B6, BY color: < BY6, R color: < R6 | |
1-vinyl-2-pyrrolidinone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, 1-vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, n-vinyl-2-pyrrolidinone, n-vinyl-2-pyrrolidone, n-vinylpyrrolidinone, n-vinylpyrrolidone, vinylpyrrolidone | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
1-ethenylpyrrolidin-2-one | |
6917 | |
Passes Test | |
Powder or Flakes | |
400ppm max. (titanyl sulphate method) | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether | |
5% Max. (K.F.) | |
3.0 to 5.0 (5% aq. soln.) |
K12 | |
0.05% max. | |
C6H9NO | |
15, 7814 | |
1ppm max. | |
10ppm max. | |
C=CN1CCCC1=O | |
111.144 | |
CHEBI:82551 | |
Authentic | |
11.5 to 12.8% (nitrogen, on anh. subst.), 10ppm 1-vinylpyrrolidin-2-one max. (HPLC), 3.0% max. 2-pyrrolidone (HPLC) | |
Plastic bottle | |
0.1% max. | |
White to White-Yellow | |
1kg |
Chemical Identifiers
9003-39-8 | |
111.144 | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
6917 | |
1-ethenylpyrrolidin-2-one |
C6H9NO | |
MFCD01076626 | |
1-vinyl-2-pyrrolidinone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, 1-vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, n-vinyl-2-pyrrolidinone, n-vinyl-2-pyrrolidone, n-vinylpyrrolidinone, n-vinylpyrrolidone, vinylpyrrolidone | |
CHEBI:82551 | |
C=CN1CCCC1=O |
Safety and Handling
LIGHT SENSITIVE
HYGROSCOPIC
RTECSNumber : TR8250000
TSCA : TSCA