missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95, Thermo Scientific Chemicals
$65.60 - $243.51
Chemical Identifiers
CAS | 9003-39-8 |
---|---|
Molecular Formula | (C6H9NO)n |
Molecular Weight (g/mol) | 111.14 |
MDL Number | MFCD01076626 |
InChI Key | WHNWPMSKXPGLAX-UHFFFAOYSA-N |
Synonym | n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone |
PubChem CID | 6917 |
ChEBI | CHEBI:82551 |
IUPAC Name | 1-ethenylpyrrolidin-2-one |
SMILES | *-CC(-*)N1CCCC1=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC227470250
|
Thermo Scientific Chemicals
227470250 |
25 g | Plastic bottle |
Each for $65.60
|
|
||||
AC227475000
|
Thermo Scientific Chemicals
227475000 |
500 g | Plastic bottle |
Each for $243.51
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9003-39-8 | |
111.14 | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
6917 | |
1-ethenylpyrrolidin-2-one |
(C6H9NO)n | |
MFCD01076626 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
CHEBI:82551 | |
*-CC(-*)N1CCCC1=O |
Specifications
Cream-White to White | |
Powder | |
(C6H9NO)n | |
15, 7814 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
1-ethenylpyrrolidin-2-one | |
6917 | |
12 to 12.8% (nitrogen) | |
Authentic | |
5% max. (K.F.) |
4 to 7 (5% aq. soln.) | |
9003-39-8 | |
MFCD01076626 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
25 g | |
*-CC(-*)N1CCCC1=O | |
111.14 | |
CHEBI:82551 | |
positive | |
Plastic bottle | |
Polyvinylpyrrolidone, Average M.W. 1.300.000, K85-95 |
RUO – Research Use Only