missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene Test Film

Catalog No. AA44502KT
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44502KT 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA44502KT Supplier Thermo Scientific Chemicals Supplier No. 044502KT
Only null left

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Quantity 1 Ea.
Chemical Name or Material Polystyrene Test Film
TSCA U
Recommended Storage Ambient temperatures

RUO – Research Use Only