Please login to your online account to display your discounted pricing

Polystyrene standard, M.W. 30,000, Mw/Mn 1.06, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Polystyrene standard
Name Note M.W. 30, 000, Mw/Mn 1.06
CAS 9003-53-6
Physical Form Powder
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4193003

  SDS
alfa aesar
4193003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4193006

  SDS
alfa aesar
4193006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Quantity 1g
Color White
Ignition Point 427°C (801°F)
CAS 9003-53-6
Odor Odorless
Chemical Name or Material Polystyrene standard
MDL Number MFCD00084450
Name Note M.W. 30, 000, Mw/Mn 1.06
Appearance White powder