Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, fluorescent, 6.0 micron, 2.5 wt% dispersion in water

Diagnostic and protein binding assays

$375.58 - $375.58

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
Name Note Fluorescent, 6.0 micron, 2.5 wt% dispersion in water
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD03703262
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA441449W

  SDS
alfa aesar™
441449W
2mL Each for $375.58
Description & Specifications

Specifications

Chemical Name or Material Polystyrene latex microsphere
TSCA TSCA
Name Note Fluorescent, 6.0 micron, 2.5 wt% dispersion in water
Physical Form Dispersion
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD03703262
CAS 9003-53-6
Odor Odorless
Recommended Storage 2°C to 8°C
Color Opalescent
Quantity 2mL
RTECS Number WL6475000
Safety and Handling

2°C to 8°C