Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, carboxylated, 0.35 micron, 2.5 wt% dispersion in water

$236.57 - $236.57

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
Name Note Carboxylated, 0.35 micron, 2.5 wt% dispersion in water
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA437174Y

  SDS
alfa aesar™
437174Y
15mL Each for $236.57
Description & Specifications

Specifications

Chemical Name or Material Polystyrene latex microsphere
Name Note Carboxylated, 0.35 micron, 2.5 wt% dispersion in water
MDL Number MFCD00084450
Odor Odorless
Recommended Storage 2°C to 8°C
Quantity 15mL
TSCA TSCA
Physical Form Dispersion
Color Opalescent
Safety and Handling

2°C to 8°C