Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 90.0 micron, 2.5 wt% dispersion in water

$284.78 - $284.78

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42751AA

  SDS
alfa aesar™
42751AA
5mL Each for $284.78
Description & Specifications

Specifications

Molecular Formula [-CH2CH(C6H5)-]n
Particle Size 90.0 Micron
MDL Number MFCD00084450
Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
Physical Form Dispersion
Quantity 5mL
Odor Odorless
Color White
TSCA TSCA
Concentration 2.5 wt% dispersion in water