Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 75.0 micron, 2.5 wt% dispersion in water

$278.61 - $278.61

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42750AA

 
alfa aesar™
42750AA
5mL Each for $278.61
Description & Specifications

Specifications

Chemical Name or Material Polystyrene latex microsphere
TSCA TSCA
Physical Form Dispersion
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD00084450
CAS 9003-53-6
Odor Odorless
Particle Size 75.0 Micron
Recommended Storage −30°C to −10°C
Color Opalescent
Concentration 2.5 wt% dispersion in water
Quantity 5mL
RTECS Number WL6475000
Safety and Handling

−30°C to −10°C