missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polystyrene latex microsphere, 75.0 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 042750AA
Description
Polystyrene is one of the most widely used plastics. Uses include protective packaging, containers, lids, bottles, trays, tumblers, disposable cutlery and produce polybutadiene rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Polystyrene latex microsphere | |
9003-53-6 | |
MFCD00084450 | |
Soluble in water. | |
PPBRXRYQALVLMV-UHFFFAOYSA-N | |
styrene | |
7501 | |
2.5 wt% dispersion in water | |
75.0 Micron |
Dispersion | |
(C8H8)n | |
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene | |
5 mL | |
*-CC(-*)C1=CC=CC=C1 | |
104.15 | |
CHEBI:27452 | |
Odorless |
Chemical Identifiers
9003-53-6 | |
104.15 | |
PPBRXRYQALVLMV-UHFFFAOYSA-N | |
7501 | |
styrene |
(C8H8)n | |
MFCD00084450 | |
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene | |
CHEBI:27452 | |
*-CC(-*)C1=CC=CC=C1 |
Safety and Handling
RTECSNumber : WL6475000
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only