Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 6.0 micron, 2.5 wt% dispersion in water

$308.89 - $308.89

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42716AA

  SDS
alfa aesar™
42716AA
5mL Each for $308.89
Description & Specifications

Specifications

Quantity 5mL
Odor Odorless
Particle Size 6.0 Micron
RTECS Number WL6475000
CAS 9003-53-6
Color Opalescent
Concentration 2.5 wt% dispersion in water
Chemical Name or Material Polystyrene latex microsphere
Storage Note 1 2°C to 8°C
Physical Form Dispersion
TSCA TSCA
MDL Number MFCD00084450
Safety and Handling

2°C to 8°C