missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polystyrene latex microsphere, 6.0 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 042716AA

Catalog No. AA42716AA



Description

Description

Polystyrene latex microsphere is used as a biocompatible material, especially in dentistry. They are used as insulators, for injection molding and casting, as sheets, plates, rods, rigid forms and beads. It is also used as extrusion molded foamed packaging, dinnerware, bottles, toys and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Polystyrene latex microsphere
9003-53-6
MFCD00084450
Fully miscible with water.
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene
7501
2.5 wt% dispersion in water
6.0 Micron
Dispersion
(C8H8)n
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
5 mL
*-CC(-*)C1=CC=CC=C1
104.15
CHEBI:27452
Odorless

Chemical Identifiers

9003-53-6
104.15
PPBRXRYQALVLMV-UHFFFAOYSA-N
7501
ethenylbenzene
(C8H8)n
MFCD00084450
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
CHEBI:27452
*-CC(-*)C1=CC=CC=C1
Safety and Handling

Safety and Handling

RTECSNumber : WL6475000

TSCA : Yes

storageNote1 : 2°C to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only