Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 45.0 micron, 2.5 wt% dispersion in water

$269.72 - $269.72

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42749AA

  SDS
alfa aesar™
42749AA
5mL Each for $269.72
Description & Specifications

Specifications

Physical Form Dispersion
Odor Odorless
Recommended Storage 2°C to 8°C
TSCA TSCA
Quantity 5mL
Particle Size 45.0 Micron
CAS 9003-53-6
RTECS Number WL6475000
Chemical Name or Material Polystyrene latex microsphere
Concentration 2.5 wt% dispersion in water
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD00084450
Color Opalescent
Safety and Handling

2°C to 8°C