Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 4.5 micron, 2.5 wt% dispersion in water

$317.93 - $317.93

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42715AA

  SDS
alfa aesar™
42715AA
5mL Each for $317.93
Description & Specifications

Specifications

Physical Form Dispersion
MDL Number MFCD00084450
Storage Note 1 2°C to 8°C
Chemical Name or Material Polystyrene latex microsphere
Concentration 2.5 wt% dispersion in water
Particle Size 4.5 Micron
Odor Odorless
CAS 9003-53-6
Color Opalescent
Quantity 5mL
RTECS Number WL6475000
TSCA TSCA
Safety and Handling

2°C to 8°C