Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 25.0 micron, 2.5 wt% dispersion in water

$349.42 - $349.42

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42730AA

 
alfa aesar™
42730AA
5mL Each for $349.42
Description & Specifications

Specifications

Chemical Name or Material Polystyrene latex microsphere
TSCA TSCA
Physical Form Dispersion
MDL Number MFCD00084450
CAS 9003-53-6
Odor Odorless
Particle Size 25.0 Micron
Storage Note 1 2°C to 8°C
Color Opalescent
Concentration 2.5 wt% dispersion in water
Quantity 5mL
RTECS Number WL6475000
Safety and Handling

2°C to 8°C