Learn More
Polystyrene latex microsphere, 20.0 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 042719AA
Description
Polystyrene latex microsphere is used as a biocompatible material, especially in dentistry. They are used as insulators, for injection molding and casting, as sheets, plates, rods, rigid forms and beads. It is also used as extrusion molded foamed packaging, dinnerware, bottles, toys and disposable cutlery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
Dispersion | |
(C8H8)n | |
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene | |
5 mL | |
*-CC(-*)C1=CC=CC=C1 | |
104.15 | |
CHEBI:27452 | |
Odorless |
Polystyrene latex microsphere | |
9003-53-6 | |
MFCD00084450 | |
Fully miscible with water. | |
PPBRXRYQALVLMV-UHFFFAOYSA-N | |
styrene | |
7501 | |
2.5 wt% dispersion in water | |
20.0 Micron |
Chemical Identifiers
9003-53-6 | |
104.15 | |
PPBRXRYQALVLMV-UHFFFAOYSA-N | |
7501 | |
styrene |
(C8H8)n | |
MFCD00084450 | |
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene | |
CHEBI:27452 | |
*-CC(-*)C1=CC=CC=C1 |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only