Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 20.0 micron, 2.5 wt% dispersion in water

$281.77 - $281.77

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42719AA

  SDS
alfa aesar™
42719AA
5mL Each for $281.77
Description & Specifications

Specifications

Physical Form Dispersion
Quantity 5mL
Particle Size 20.0 Micron
Odor Odorless
Color White
Chemical Name or Material Polystyrene latex microsphere
TSCA TSCA
CAS 9003-53-6
MDL Number MFCD00084450
Concentration 2.5 wt% dispersion in water