Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 2 micron, 2.5 wt% dispersion in water

$257.05 - $257.05

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA427444Y

  SDS
alfa aesar™
427444Y
15mL Each for $257.05
Description & Specifications

Specifications

Chemical Name or Material Polystyrene latex microsphere
TSCA TSCA
Physical Form Dispersion
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD00084450
CAS 9003-53-6
Odor Odorless
Particle Size 2 Micron
Recommended Storage 2°C to 8°C
Color Opalescent
Concentration 2.5 wt% dispersion in water
Quantity 15mL
RTECS Number WL6475000
Safety and Handling

2°C to 8°C