Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 15.0 micron, 2.5 wt% dispersion in water

$339.03 - $339.03

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42718AA

  SDS
alfa aesar™
42718AA
5mL Each for $339.03
Description & Specifications

Specifications

Physical Form Dispersion
Odor Odorless
TSCA TSCA
Storage Note 1 2°C to 8°C
Quantity 5mL
Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
Concentration 2.5 wt% dispersion in water
RTECS Number WL6475000
MDL Number MFCD00084450
Particle Size 15.0 Micron
Color Opalescent
Safety and Handling

2°C to 8°C