Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 10.0 micron, 2.5 wt% dispersion in water

$303.24 - $303.24

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42717AA

 
alfa aesar™
42717AA
5mL Each for $303.24
Description & Specifications

Specifications

Chemical Name or Material Polystyrene latex microsphere
TSCA TSCA
Physical Form Dispersion
MDL Number MFCD00084450
CAS 9003-53-6
Odor Odorless
Particle Size 10.0 Micron
Storage Note 1 2°C to 8°C
Color Opalescent
Concentration 2.5 wt% dispersion in water
Quantity 5mL
RTECS Number WL6475000
Safety and Handling

2°C to 8°C